2-(9,9'-Spirobi[fluoren]-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:884336-44-1
:
Formula:C₃₁H₂₇BO₂
MDL Number:MFCD29472553
:442.37 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:248.0 to 252.0 deg-C
Specification Test Results
Frequently Bought Together