(S)-2,2',3,3'-Tetrahydro-1,1'-spirobi[1H-indene]-7,7'-diol ≥98.0% (by GC)
Catalog # TCIAS0958-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystaline
- Purity (GC):min. 98.0 %
- Optical purity (LC):min. 98.0 ee%
- Melting point:156.0 to 160.0 deg-C
Specification Test Results
About this item
:
:252.31 g/mol
CAS Number:223259-63-0
:MFCD16621090
Formula:C₁₇H₁₆O₂