
LC-SMCC (N-Succinimidyl 6-[[4-(N-Maleimidomethyl)cyclohexyl]carboxamido]hexanoate) ≥98.0% (by HPLC)
Catalog # TCIAS0881-25MG
Supplier: TCI
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:180.0 to 184.0 deg-C
Specification Test Results
About this item
:
:447.49 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:125559-00-4
Formula:C₂₂H₂₉N₃O₇
MDL Number:MFCD01863459