Order Entry
Netherlands
ContactUsLinkComponent
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate) ≥97.0% (by HPLC)
Irritant
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate) ≥97.0% (by HPLC)
Catalog # TCIAS0861-10MG
Supplier:  TCI
CAS Number:  
Irritant
SMPB (N-Succinimidyl 4-(4-Maleimidophenyl)butyrate) ≥97.0% (by HPLC)
Catalog # TCIAS0861-10MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Melting point:
    129.0 to 133.0 deg-C

Specification Test Results

About this item

 :
 :356.33 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:79886-55-8
Formula:C₁₈H₁₆N₂O₆
MDL Number:MFCD00054961