Order Entry
Netherlands
ContactUsLinkComponent
SIA (N-Succinimidyl iodoacetate) ≥95.0% (by total nitrogen basis)
Irritant
SIA (N-Succinimidyl iodoacetate) ≥95.0% (by total nitrogen basis)
Catalog # TCIAS0844-100MG
Supplier:  TCI
CAS Number:  
Irritant
SIA (N-Succinimidyl iodoacetate) ≥95.0% (by total nitrogen basis)
Catalog # TCIAS0844-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (with Total Nitrogen):
    min. 95.0 %
  • Melting point:
    146.0 to 150.0 deg-C

Specification Test Results

About this item

 :
 :283.02 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:39028-27-8
Formula:C₆H₆INO₄
MDL Number:MFCD00058451