Sodium sulphanilate hydrate ≥98.0% (by titrimetric analysis) for biochemical research
Catalogus # TCIAS0832-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Ion exchange titration):min. 98.0 % (calcd.on anh.substance)
- Water:max. 16.0 %
- Nitrite ion detection test (Griess method):to pass test
Specification Test Results
About this item
:
:195.17 g/mol
Merck Index:14,08926
CAS Number:123333-70-0
Formula:C₆H₇NO₃S
MDL Number:MFCD00064394