N-Succinimidyl 3-(Diphenylphosphino)propionate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:170278-50-9
:
Formula:C₁₉H₁₈NO₄P
:355.33 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Frequently Bought Together