Order Entry
Netherlands
ContactUsLinkComponent
Scandium(III) acetate hydrate ≥98.0% (by titrimetric analysis)
Scandium(III) acetate hydrate ≥98.0% (by titrimetric analysis)
  TCIAS0534-1G
 :  TCI
 :  
Scandium(III) acetate hydrate ≥98.0% (by titrimetric analysis)
  TCIAS0534-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 % (calcd.on anh.substance)
  • Water:
    max. 3.0 %

 

 

 :
 :240.10 g/mol
 :223-274-5
 :304675-64-7
 :207671-51-0
 :C₆H₉O₆Sc·H₂O
 :MFCD00150122