Succinimidyl-6-[[7-(N,N-dimethylaminosulphonyl)-2,1,3-benzoxadiazol-4-yl]amino]hexanoate ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAS0503-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:137.0 to 141.0 deg-C
Specification Test Results
About this item
:
:453.47 g/mol
Storage Temperature:Refrigerator
CAS Number:1858255-08-9
Formula:C₁₈H₂₃N₅O₇S