N-Succinimidyl 6-Biotinamidohexanoate ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:72040-63-2
:
Formula:C₂₀H₃₀N₄O₆S
MDL Number:MFCD00065502
:454.54 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Elemental analysis (Nitrogen):11.60 to 12.80 %
Specification Test Results
Frequently Bought Together