
DMS (Dimethyl suberimidate dihydrochloride) ≥98.0% (by titrimetric analysis) cross-linking agent, for protein research
Supplier: TCI
Certificates
About this item
CAS Number:34490-86-3
:
Formula:C₁₀H₂₀N₂O₂
MDL Number:MFCD00012574
:273.20 g/mol
EINECS:252-060-4
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together