Scopolamine methyl bromide ≥98.0% (by titrimetric analysis)
Catalog # TCIAS0231-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Specific rotation [a]20/D:-22.0 to -25.0 deg (C=1, H2O)
- Solubility in Water:almost transparency
Specification Test Results
About this item
:
:398.30 g/mol
Merck Index:14,06003
EINECS:205-844-5
UN:1544
ADR:6.1,III
CAS Number:155-41-9
Formula:C₁₈H₂₄BrNO₄
MDL Number:MFCD00078560