Order Entry
Netherlands
ContactUsLinkComponent
Sinigrin hydrate ≥98.0% (by HPLC)
Sinigrin hydrate ≥98.0% (by HPLC)
Catalog # TCIAS0156-100MG
Supplier:  TCI
CAS Number:  
Sinigrin hydrate ≥98.0% (by HPLC)
Catalog # TCIAS0156-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Specific rotation [a]20/D:
    -15.0 to -20.0 deg (C=1, H2O)
  • Water:
    2.0 to 7.0 %

Specification Test Results

About this item

 :
 :397.45 g/mol
Merck Index:14,08545
EINECS:223-545-8
CAS Number:3952-98-5
Formula:C₁₀H₁₇NO₉S₂
MDL Number:MFCD00006616