Diacetato{(R)-(+)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}ruthenium(II)
Supplier: TCI
Certificates
About this item
CAS Number:374067-50-2
:
MDL Number:MFCD09753022
Storage Temperature:Refrigerator
Specification Test Results
- IR:to pass test
Specification Test Results
Frequently Bought Together