RuCl2[(S)-dm-segphos®][(S,S)-dpen]
TCIAR0130-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
- Appearance:Light yellow to Amber to Dark green powder to crystal
- IR:to pass test
: Dichloro{(S)-(-)-5,5'-bis[di(3,5-xylyl)phosphino]-4,4'-bi-1,3-benzodioxole}[(1S,2S)-(-)-1,2-diphenylethylenediamine]ruthenium(II) RuCl2[(S)-dm-segphos®][(S,S)-dpen]
:
:1107.07 g/mol
:944450-46-8
:C₆₀H₆₀Cl₂N₂O₄P₂Ru
:MFCD09753032