Order Entry
Netherlands
ContactUsLinkComponent
RuCl[(R,R)-Tsdpen](mesitylene) ≥90% (by HPLC)
RuCl[(R,R)-Tsdpen](mesitylene) ≥90% (by HPLC)
Supplier:  TCI
Certificates
undefined
RuCl[(R,R)-Tsdpen](mesitylene) ≥90% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (HPLC):
    min. 90 area%

Specification Test Results

Related Information

Frequently Bought Together