β-L-Ribofuranose-1-acetate 2,3,5-tribenzoate ≥98.0% (by HPLC)
Catalog # TCIAR0080-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:128.0 to 132.0 deg-C
- Specific rotation [a]20/D:-22.0 to -26.0 deg (C=1, Pyridine)
Specification Test Results
About this item
:
:504.49 g/mol
CAS Number:3080-30-6
:MFCD00628242
Formula:C₂₈H₂₄O₉
MDL Number:MFCD04114304