Ranitidine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:66357-59-3
:
Formula:C₁₃H₂₂N₄O₃S
MDL Number:MFCD00069339
:350.86 g/mol
Merck Index:14,08110
Storage Temperature:Refrigerator
EINECS:266-333-0
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together