Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
1-(1-Adamantyl)ethylamine hydrochloride ≥98.0% (by total nitrogen basis)
Catalog # TCIAR0070-5G
Supplier:  TCI
CAS Number:  
Irritant
1-(1-Adamantyl)ethylamine hydrochloride ≥98.0% (by total nitrogen basis)
Catalog # TCIAR0070-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Solubility in hot Water:
    almost transparency

Specification Test Results

About this item

 :
 :215.77 g/mol
Merck Index:14,08224
Melting Pt:375°C
 :GHS07
CAS Number:1501-84-4
Formula:C₁₂H₂₁N
MDL Number:MFCD00072023