1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:6974-32-9
:
Formula:C₂₈H₂₄O₉
MDL Number:MFCD00005357
:504.5 g/mol
EINECS:230-220-4
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:127.0 to 132.0 deg-C
- Specific rotation [a]20/D:+22.0 to +26.0 deg (C=1, Pyridene)
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together