Order Entry
Netherlands
ContactUsLinkComponent
Rutin hydrate ≥94.0% (by HPLC, titration analysis)
Rutin hydrate ≥94.0% (by HPLC, titration analysis)
Catalog # TCIAR0035-25G
Supplier:  TCI
CAS Number:  
Rutin hydrate ≥94.0% (by HPLC, titration analysis)
Catalog # TCIAR0035-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 94.0 area%
  • Purity (Neutralization titration):
    min. 94.0 % (calcd.on anh.substance)
  • Water:
    6.0 to 8.5 %

Specification Test Results

About this item

 :
 :610.52 g/mol
Merck Index:14,08304
EINECS:205-814-1
CAS Number:207671-50-9
Formula:C₂₇H₃₀O₁₆
MDL Number:MFCD00006830