Order Entry
Netherlands
ContactUsLinkComponent
Quinacridone ≥99.0% (by total nitrogen basis), purified by sublimation
Quinacridone ≥99.0% (by total nitrogen basis), purified by sublimation
Catalog # TCIAQ0083-1G
Supplier:  TCI
CAS Number:  
Quinacridone ≥99.0% (by total nitrogen basis), purified by sublimation
Catalog # TCIAQ0083-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Purity (with Total Nitrogen):
    min. 99.0 %

Specification Test Results

About this item

 :
 :312.33 g/mol
Melting Pt:390°C
EINECS:213-879-2
CAS Number:1047-16-1
Formula:C₂₀H₁₂N₂O₂
MDL Number:MFCD00059956