Polyethylene glycol methyl ether methacrylate n=∼9 stabilised
Supplier: TCI
Certificates
About this item
CAS Number:26915-72-0
:
Formula:CH₃(OCH₂CH₂)nOCOC(=CH₂)CH₃
MDL Number:MFCD00241432
:Stabilised with 4-methoxyphenol (MEHQ)
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- The average value of n:8.5 to 9.5
Specification Test Results
Frequently Bought Together