[6,6]-Phenyl-C71-butyric acid methyl ester (mixture of isomers) mixture of isomers ≥99.0% (by HPLC) for organic electronics
Catalog # TCIAP2683-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Black powder to crystal
- Purity (HPLC):min. 99.0 % (total of isomer)
- Absorbance (E1%1cm):min. 250 (Toluene, 370.0 to 375.0nm)
- Absorbance (E1%1cm)2:min. 200 (Toluene, 460.0 to 465.0nm)
- Electron Mobility (muFET):min 0.015 cm2/Vs (HMDS Si/SiO2 substrate)
Specification Test Results
About this item
:
:1031.01 g/mol
CAS Number:609771-63-3
Formula:C₈₂H₁₄O₂
MDL Number:MFCD09842750