
4-(2-Pyridyl)benzonitrile ≥97.0% (by GC)
TCIAP2659-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:98.0 to 102.0 deg-C
:
:180.21 g/mol
:250-921-9
:GHS07
:32111-34-5
:C₁₂H₈N₂
:MFCD04116306