Pregna-4,6-diene-3,20-dione ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1162-56-7
:
Formula:C₂₁H₂₈O₂
MDL Number:MFCD00199858
:312.45 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:144.0 to 148.0 deg-C
- Specific rotation [a]20/D:140 to 155 deg (C=1, EtOH)
Specification Test Results
Frequently Bought Together