
9,9'-Spirobi[9H-fluoren]-2-amine ≥98.0% (by HPLC, titration analysis)
Catalogus # TCIAP2585-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Green to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:231.0 to 235.0 deg-C
Specification Test Results
About this item
:
:331.42 g/mol
:GHS07
CAS Number:118951-68-1
Formula:C₂₅H₁₇N
MDL Number:MFCD27956479