
9-Phenyl-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole ≥96.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:1035631-57-2
:
Formula:C₃₀H₃₅B₂NO₄
:495.23 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (with Total Nitrogen):min. 96.0 %
- Melting point:235.0 to 240.0 deg-C
Specification Test Results
Frequently Bought Together