Poly(2-propyl-2-oxazoline) n=∼100, ULTROXA®
TCIAP2508-200MG
: TCI
:
- Pk:200 mg
- Seq:2530
- Appearance:White to Light yellow powder to crystal
- Average molecular weight:9,200 to 13,800
- Molecular weight distribution:max. 1.20
:
:Refrigerator
:941228-32-6