

1-(2-Cyanophenyl)piperazine ≥98.0% (by GC, titration analysis)
Catalog # TCIAP2448-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
:187.25 g/mol
Boiling Pt:292°C
Flash Pt:47°C
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:111373-03-6
Formula:C₁₁H₁₃N₃
MDL Number:MFCD00040793