Order Entry
Netherlands
ContactUsLinkComponent
 
1,2,3,3,8-Pentamethyl-5-(trifluoromethyl)-2,3-dihydro-1H-pyrrolo[3,2-g]quinolin-7(8H)-one ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAP2441-200MG
Supplier:  TCI
CAS Number:  
1,2,3,3,8-Pentamethyl-5-(trifluoromethyl)-2,3-dihydro-1H-pyrrolo[3,2-g]quinolin-7(8H)-one ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAP2441-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Molar extinction coefficient:
    min. 21500 (EtOH, 385 to 390nm)

Specification Test Results

About this item

 :
 :324.35 g/mol
EINECS:261-404-2
CAS Number:58721-74-7
Formula:C₁₇H₁₉F₃N₂O
MDL Number:MFCD00467063