trans-4-n-Propylcyclohexyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:83242-83-5
:
Formula:C₂₅H₄₄O₂
MDL Number:MFCD20527218
:376.62 g/mol
EINECS:280-293-1
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Mesomorphic range:151.0 to 190.0 deg-C
Specification Test Results
Frequently Bought Together