
Phenyl[3-(trifluoromethyl)phenyl]iodonium trifluoromethanesulphonate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:905718-46-9
:
Formula:C₁₄H₉F₆IO₃S
MDL Number:MFCD21608481
:498.18 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 95.0 %
- Melting point:110.0 to 114.0 deg-C
Specification Test Results
Frequently Bought Together