(S)-[4-(Pyridin-4-yl)-4,5-dihydro-3H-dinaphtho[2,1-c:1',2'-e]azepine-2,6-diyl]bis[bis[4-(tert-butyl)phenyl]methanol] ≥85.0% (by HPLC)
Catalog # TCIAP2380-50MG
Supplier: TCI
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 85.0 area%
- Elemental analysis (Nitrogen):2.50 to 3.30 %
- Specific rotation [a]20/D:-270 to -310 deg (C=0.2, CHCl3)
Specification Test Results
About this item
:
:961.35 g/mol
CAS Number:1883396-49-3
Formula:C₆₉H₇₂N₂O₂