4-[2-(trans-4-Pentylcyclohexyl)ethyl]cyclohexanone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:121040-08-2
:
Formula:C₁₉H₃₄O
MDL Number:MFCD14636435
:278.48 g/mol
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Melting point:42.0 to 46.0 deg-C
Specification Test Results
Frequently Bought Together