
4-(Pentyloxy)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:146449-90-3
:
Formula:C₁₁H₁₇BO₃
MDL Number:MFCD04039035
:208.06 g/mol
Melting Pt:80°C
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Neutralization titration):97.0 to 110.0 %
Specification Test Results
Frequently Bought Together