Order Entry
Netherlands
ContactUsLinkComponent
 
 
2-(2-Propynyl)-1H-isoindole-1,3(2H)-dione ≥98.0% (by GC)
 :  TCI
 
undefined
 
2-(2-Propynyl)-1H-isoindole-1,3(2H)-dione ≥98.0% (by GC)
 :  TCI

 

  • Purity (GC):
    min. 98.0 %
  • Melting point:
    151.0 to 155.0 deg-C

 

Frequently Bought Together