Order Entry
Netherlands
ContactUsLinkComponent
8-[2-(2-Pyridyl)phenyl]-5H-pyrido[3,2-b]indole ≥98.0% (by GC)
8-[2-(2-Pyridyl)phenyl]-5H-pyrido[3,2-b]indole ≥98.0% (by GC)
Catalog # TCIAP2307-100MG
Supplier:  TCI
CAS Number:  
undefined
8-[2-(2-Pyridyl)phenyl]-5H-pyrido[3,2-b]indole ≥98.0% (by GC)
Catalog # TCIAP2307-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :321.38 g/mol
CAS Number:1251503-35-1
Formula:C₂₂H₁₅N₃