
Pentafluorophenyl methacrylate ≥97.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:13642-97-2
:
Formula:C₁₀H₅F₅O₂
MDL Number:MFCD00042332
:252.14 g/mol
Flash Pt:77°C
:Stabilised with 4-methoxyphenol (MEHQ)
:GHS07
Storage Temperature:Koelkast
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together