
(Piperidinium-1-ylmethyl)trifluoroborate ≥96.0% (by total nitrogen basis)
Catalog # TCIAP2287-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Purity (with Total Nitrogen):min. 96.0 %
Specification Test Results
About this item
:
:166.98 g/mol
Melting Pt:150°C
UN:1759
ADR:8,III
:GHS05
CAS Number:1268340-93-7
Formula:C₆H₁₃BF₃N
MDL Number:MFCD18071160