Order Entry
Netherlands
ContactUsLinkComponent
5H-Pyrido[3,2-b]indole ≥98.0% (by HPLC)
5H-Pyrido[3,2-b]indole ≥98.0% (by HPLC)
Catalog # TCIAP2274-100MG
Supplier:  TCI
CAS Number:  
undefined
5H-Pyrido[3,2-b]indole ≥98.0% (by HPLC)
Catalog # TCIAP2274-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Nitrogen):
    16.00 to 17.50 %
  • Melting point:
    211.0 to 215.0 deg-C

Specification Test Results

About this item

 :
 :168.20 g/mol
CAS Number:245-08-9
Formula:C₁₁H₈N₂
MDL Number:MFCD13178683