Order Entry
Netherlands
ContactUsLinkComponent
4'-(4-Pyridyl)-2,2':6',2''-terpyridine ≥97.0% (by total nitrogen basis)
Irritant
4'-(4-Pyridyl)-2,2':6',2''-terpyridine ≥97.0% (by total nitrogen basis)
Catalog # TCIAP2202-200MG
Supplier:  TCI
CAS Number:  
undefined
Irritant
4'-(4-Pyridyl)-2,2':6',2''-terpyridine ≥97.0% (by total nitrogen basis)
Catalog # TCIAP2202-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Purity (HPLC):
    min. 95.0 area%
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    221.0 to 226.0 deg-C

Specification Test Results

About this item

 :
 :310.36 g/mol
 :GHS07
CAS Number:112881-51-3
Formula:C₂₀H₁₄N₄
MDL Number:MFCD00932037