Polyethylene glycol monomethyl ether 4000
Supplier: TCI
Certificates
About this item
CAS Number:9004-74-4
:
Formula:CH₃(OCH₂CH₂)nOH
MDL Number:MFCD00084416
Specification Test Results
- Appearance:White to Almost white powder to lump
- Average molecular weight:3,800 to 4,200
- Hydroxyl Value:13.0 to 15.0
- Melting point:58.0 to 64.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together