Polyethylene glycol monomethyl ether 2000
Supplier: TCI
Certificates
About this item
CAS Number:9004-74-4
:
Formula:CH₃(OCH₂CH₂)nOH
MDL Number:MFCD00084416
Specification Test Results
- Appearance:White to Almost white powder to crystaline
- Average molecular weight:1,900 to 2,100
- Hydroxyl Value:25.0 to 30.0
- Melting point:49.0 to 55.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together