Polyethylene glycol monomethyl ether 1000
Supplier: TCI
Certificates
About this item
CAS Number:9004-74-4
:
Formula:CH₃(OCH₂CH₂)nOH
MDL Number:MFCD00084416
Specification Test Results
- Appearance:White to Almost white powder to lump
- Average molecular weight:950 to 1,050
- Hydroxyl Value:50.0 to 60.0
Specification Test Results
Related Information
Frequently Bought Together