
Potassium[[(tert-butoxycarbonyl)amino]methyl]trifluoroborate ≥97.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:1314538-55-0
:
Formula:C₆H₁₂BF₃KNO₂
MDL Number:MFCD19686142
:237.07 g/mol
Melting Pt:181°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 97.0 %
Specification Test Results
Frequently Bought Together