Order Entry
Netherlands
ContactUsLinkComponent
2-(2-Pyridyl)indole ≥97.0% (by GC)
2-(2-Pyridyl)indole ≥97.0% (by GC)
Catalog # TCIAP2142-200MG
Supplier:  TCI
CAS Number:  
undefined
2-(2-Pyridyl)indole ≥97.0% (by GC)
Catalog # TCIAP2142-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Green powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    152.0 to 156.0 deg-C

Specification Test Results

About this item

 :
 :194.24 g/mol
CAS Number:13228-40-5
Formula:C₁₃H₁₀N₂
MDL Number:MFCD00033466