3-(4-Pyridyl)-L-alanine dihydrochloride ≥96.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:178933-04-5
:
Formula:C₈H₁₀N₂O₂·2HCl
MDL Number:MFCD13184932
:239.1 g/mol
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (with Total Nitrogen):min. 96.0 %
- Specific rotation [a]20/D:+23.0 to +26.0 deg (C=2,H2O)
Specification Test Results
Frequently Bought Together