Pigment Orange 43 ≥94.0% (by total nitrogen basis)
Catalog # TCIAP2124-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Orange to Brown powder to crystal
- Purity (with Total Nitrogen):min. 94.0 %
Specification Test Results
About this item
:
:412.41 g/mol
EINECS:224-597-4
CAS Number:4424-06-0
Formula:C₂₆H₁₂N₄O₂
MDL Number:MFCD00071829