PTCBI (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by total nitrogen basis)
Catalog # TCIAP2119-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Black powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
:
:536.55 g/mol
CAS Number:79534-91-1
Formula:C₃₆H₁₆N₄O₂